Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 56-910 PVC Coat Apron

Catalog No. 19010247 Shop All Ansell Products
Change view
Click to view available options
Size:
3X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-247 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
N/A
Catalog No. 19-010-247 Supplier Ansell Supplier No. 952275
  • $986.66
    $432.65 / Case of 12

    Save $554.01

    56% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
N/A

Medium to heavy duty splash protection coat aprons offers greater body coverage.

  • Reliable protection against oil, grease, fats, chemicals, abrasion, punctures
  • No exposed elastic or metal parts to cause static or corrosion
  • Protection where splashes from acids and caustic bases are a concern
  • Virgin vinyl construction contains no reprocessed materials and is flexible
  • Sleeves come with elasticized cuffs for added protection
  • PVC (vinyl) grommets are permanently sealed to the aprons for durability and allow for size adjustment without tear-out
  • Adjustable ties ensure optimal fit for a wide range of wearers
TRUSTED_SUSTAINABILITY

Specifications

Type Apron
Size 3X-Large
Material PVC
Color Blue
Disposable Reusable
Closure Type Ties
Sterility Non-sterile
Construction Virgin Vinyl, No Reprocessed Materials, Dielectrically Sealed
Finish Dielectrically Sealed
Latex Free No
No. of Pockets None
Resistance Puncture, Abrasion, Chemicals, Fats, Oils, Grease
Thickness 8 mil
For Use With (Application) Food Service, Maintenance, Transferring Liquids
No. per Case 12/Pk.
No. per Pack 1 Ea.
Seam Type Seamless
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

N/A

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.