Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 37-510 High Performance Nitrile Gloves

Catalog No. 19501034 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-034 12
19-501-029 7
19-501-030 8
19-501-031 9
19-501-032 10
19-501-033 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-501-034 Supplier Ansell Supplier No. 184699
This item is not returnable. View return policy

Thick 11 mil nitrile gloves with UltraGrip™ technology offer an unmatched grip in fatty or wet conditions. Often used in agriculture, food processing and general industries.

  • High performance nitrile compound for enhanced flexibility and dexterity, even in cold environments
  • Secure grip on wet and dry surfaces
  • Unflocked to prevent the risk of food contamination
  • Ideal for food processing industry
  • Also can be used in the agriculture, viticulture and general industries
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, Category III
Chlorinated Yes
Color Blue
Latex Free Yes
Material Nitrile
Size 12
Thickness (English) 11 mil
Thickness (Metric) 0.28 mm
EN388 Standard EN 388:2001X, EN 388:2001
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Straight
Grip Design Raised Diamond
Lining Unlined
Model 184699
Series AlphaTech™ 37-510
Length (English) 12.8 in.
Length (Metric) 32.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.