Learn More
TARGETMOL CHEMICALS INC ALPHAVBETA1 INTEGRIN-IN- 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T13473
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (alpha)v(beta)1 integrin-IN-1 is a potent and selective inhibitor of (alpha)v(beta)1 integrin (IC50 of 0.63 nM) with antifibrotic effects. purity: 99%
Catalog No. 50-258-1344
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.