Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alprostadil, USP, 95-105%, Spectrum™ Chemical
SDP

Catalog No. 18600453 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
1 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-453 1 g Amber Glass Bottle
18-600-450 10 mg Amber Glass Bottle
18-600-451 25 mg Amber Glass Bottle
18-600-452 100 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-453 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P16791GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

P1679, 745-65-3, C20H34O5

Spectrum™ Chemical Alprostadil, USP is used to treat male sexual function problems (erectile dysfunction). All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 745-65-3
Molecular Formula C20H34O5
Molecular Weight (g/mol) 354.49
InChI Key GMVPRGQOIOIIMI-DWKJAMRDSA-N
IUPAC Name 7-[(1R,2R,3R)-3-hydroxy-2-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-5-oxocyclopentyl]heptanoic acid
SMILES CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Packaging Amber Glass Bottle
Quantity 1 g
UN Number UN2811
Percent Purity 95 to 105%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Alprostadil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.