accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OH(R)(R)HOO(R)OHO(E)

CAS RN 745-65-3

IUPAC Name: 7-[(1R,2R,3R)-3-hydroxy-2-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-5-oxocyclopentyl]heptanoic acid
Synonyms: alprostadil l-prostaglandin E1 befar Alprox-TD alprostadilum prostaglandin E1 Muse Edex caverject femprox
Molecular Weight (g/mol): 354.49
Molecular Formula: C20H34O5
InChi Key: GMVPRGQOIOIIMI-DWKJAMRDSA-N
SMILES: CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)
  • (1)
Grade 
  • (7)
Physical Form 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (7)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (7)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)