Learn More
BioVendor, LLC Acid Labile Subunit (ALS) Human ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC E35
Encompass
Acid Labile Subunit (ALS) Human ELISA (96 wells). Enzyme immunoassay kit to measure Acid Labile Subunit (ALS) in human serum or plasma. Quality controls included. SAMPLE REQ: 10 l/well. CALIBRATION RANGE: 0 200 ng/mL. ASSAY RANGE: 0.53 30 000 ng/mL. DETECTION LIMIT: 0.53 ng/ml. SUMMARY: The acid-labile subunit is an important part of the IGF-storage mechanism in circulation. In ALS deficiency or in ALS knock-out mice the concentrations of IGF and IGFBP-3 in the circulation are significantly decreased thus resulting in impaired growth. In primary ALS deficiency hyperinsulinemia could be observed. Further, the ALS-IGF-IGFBP-system seems to be of relevance in coronary disease. ALS measurement can be used as supplementary info in growth hormone studies together with IGF-I and IGFBP-3 measurements. Thus, ALS quantification is useful in studying GH-deficiency and excess. NAMES: Insulin-like growth factor-binding protein complex ALS, IGFALS. Manufactured by Mediagnost GmbH, Germany.
Catalog No. 50-417-1
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.