Learn More
Hycult Biotech Inc ALT COMP PATH MS 1X96 DET
SDP
Supplier:  Hycult Biotech Inc HIT422
Encompass
The HIT422 measures the alternative pathway activity mediated via LPS. Complement deficiencies or other defects in the complement system can easily be screened by running an assay for each pathway in parallel or separately. HYCULT BIOTECH INC mfg# HIT422
Catalog No. NC1915563
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.