Learn More
Restek™ Alta-Robbins Sample Cylinder Valves
SDP
Catalog No. 06729002 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
Sulfinert treated
Pressure:
3500 psi
5000 psi
Thread Size (Metric):
0.63 NPTM x 0.63 Male compressioncm
0.63 NPTMcm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Pressure Thread Size (Metric)
06-729-002 Sulfinert treated 3500 psi 0.63 NPTMcm
06-729-004 Sulfinert treated 3500 psi 0.63 NPTM x 0.63 Male compressioncm
06-729-010 Stainless Steel 5000 psi 0.63 NPTMcm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-729-002 Supplier Restek™ Supplier No. 21400
Add to Cart
Add to Cart
Large, durable, Kel-F seat ensures leak-free operation.
  • Temperature range: -40° to +120°C
  • Available stainless-steel or sulfinert-treated
Specifications
Includes Dip tube and rupture disks
Material Sulfinert treated
Pressure 3500 psi
Thread Size (English) 0.25 NPTM in.
Thread Size (Metric) 0.63 NPTMcm
Type Sample Cylinder Valve