Learn More
AdipoGen Altenusin (1 mg)
SDP
Supplier:  AdipoGen AGCN20143M001
Encompass_Preferred
Chemical. CAS: 31186-12-6. Formula: C15H14O6. MW: 290.3. Isolated from Penicillium sp. Antibiotic. Antifungal penicillide. Non-competitive, specific neutral sphingomyelinase (N-SMase) inhibitor. Strong pp60c-Src inhibitor. Inhibits cFMS receptor tyrosine kinase (CSF-1/m-CSF receptor tyrosine kinase) which is implicated in cancer and bone diseases. Myosin light chain kinase inhibitor. Exhibits anti-HIV-1 integrase activity. Cytotoxic activity against mouse lymphoma cells. Anti-protozoan, trypanothione reductase (TR) inhibitor. Phytotoxin.
Catalog No. 50-205-3522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.