Learn More
JDM LAB SOLUTIONS LLC AlteRead 1D barcode reader - SBS
Supplier:  JDM LAB SOLUTIONS LLC 251050
Encompass
For use with AlteRead SBS rack 2D barcode camera reader. Includes linear barcode reader and mounting bracket
Catalog No. 50-272-0023