Learn More
JDM LAB SOLUTIONS LLC AlteRead Large format & SBS rack 2D barcode camera reader
Supplier:  JDM LAB SOLUTIONS LLC 254000
Encompass
Fast 2D barcode camera rack reader. Decodes large format racks 81, 100 and all SBS formats. Decodes embedded 2D rack IDs s. Requires: 2GB RAM, Windows 10. Includes: AlteRead software. Rack templates sold separately. Export data as XML, Excel, text (.csv or .txt), JSON or image files. Warranty: 5 years
Catalog No. 50-272-0016
Only null left
Add to Cart
Request a Quote