Learn More
JDM LAB SOLUTIONS LLC AlteRead™ Set Up Pack - SBS rack format Store and track samples easily with all the kit supplied in one cost saving package
Supplier:  JDM LAB SOLUTIONS LLC 253001
Encompass
AlteRead™ Set Up Pack Includes: AlteRead Single tube 2D barcode camera reader, AlteRead SBS rack 2D barcode camera reader, 2D barcoded SBS format AlteTubes (2 cases) - choice of 0.5ml, 0.75ml, 1.1ml or 1.4ml. Tubes, racks and caps in any packaging format.
Catalog No. 50-272-0018
Only null left
Add to Cart
Request a Quote