Learn More
Altitrak
SDPSCI_ED
Catalog No. S41861 Shop All Estes Industries Products
Click to view available options
Includes:
Meters-to-feet conversion table
Catalog No. Includes
S41861 Meters-to-feet conversion table
1 options
Catalog No. S41861 Supplier Estes Industries Supplier No. 2232
Includes
Meters-to-feet conversion table
Measure the maximum altitude of your rocket.
  • Gathers experimental performance and altitude data
  • Tracks rocket in flight with easy-to-use sights
  • Follow rocket to its apogee, then lock trigger to the reading
  • Instantly and accurately determines peak altitude
  • Displays altitude in meters and elevation angle in degrees
Specifications
For Use With (Application) Gathers performance and altitude data of model rockets
Includes Meters-to-feet conversion table
Product Type Altitrak Altimeter