Learn More
Labrepco Inc Aluminum Canister, 250 mL Capacity, Sand Blasted Finish, For Blood Bag Storage
SDP
Supplier:  Labrepco Inc CZC021
Encompass_Preferred
The ALUM CANISTER FOR BXTR FEN is a robust aluminum canister designed for secure storage of 250 mL blood bags. Its sandblasted finish and clear window enhance identification and workflow efficiency in laboratory settings. Offering customization options like laser engraving and QR codes, this canister ensures precise tracking of products, helping streamline inventory management and compliance.
Catalog No. NC2204126
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.