Learn More
Description
- MP EcoChrom™ Alumina N, Activity Super I shows a sorbtion capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
- Their inital activity is kept in a very narrow range and their dactivation behavior is extremely constant.
Specifications
Specifications
| Color | Pink to White |
| Physical Form | Free-flowing Powder |
| Quantity | 1 kg |
| IUPAC Name | dialuminium(3+) trioxidandiide |
| Formula Weight | 101.96 |
| Density | 3.94 g/cm3 (at 20°C) |
| Chemical Name or Material | Alumina B, Activity: Super I |
Safety and Handling
| EINECSNumber | 215-691-6 |
| Recommended Storage | Store at room temperature. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.