Learn More
Electron Microscopy Sciences Alumina Powder Gamma, Type DX, 0.05 µm Particles 1 LB
SDP
Supplier:  Electron Microscopy Sciences 5036101
Encompass_Preferred
EMS's Alumina Powders are accurately controlled for all specs and particle size distribution, assuring you the best polishing results. Typical applications are precision optics, metallographic sections, electro-optical crystals, silicone wafers, ferrite components, gem stones, and acrylic lenses. Type DX Alumina Powder is specially treated to reduce the number of agglomerates.
Particles: 0.05 µm
Catalog No. 50-997-300
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.