Learn More
Perkin Elmer US LLC Alumina Sample Pans and Covers, Pkg. 6
Supplier:  Perkin Elmer US LLC N5190180
Encompass_Preferred
These alumina sample pans and covers are for use with DSC 4000, DSC 6000, DSC 8000, DSC 8500, Diamond DSC, Pyris 1 DSC, DSC 7, DSC 6, Pyris 6 DSC and Jade DSC.
Catalog No. 50-211-1988
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.