Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum acrylate

Catalog No. p-7051802
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4200306 5 g
AA4200314 25 g
AA4200322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4200306 Supplier Thermo Scientific Chemicals Supplier No. 04200306
Only null left
Add to Cart

CAS: 15743-20-1 | C9H12AlO6 | 243.171 g/mol

Aluminum acrylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15743-20-1
Molecular Formula C9H12AlO6
Molecular Weight (g/mol) 243.171
MDL Number MFCD00078421
InChI Key UNGWYUCJMHZVEX-UHFFFAOYSA-N
Synonym aluminum acrylate
PubChem CID 131874419
IUPAC Name aluminum;prop-2-enoic acid
SMILES C=CC(=O)O.C=CC(=O)O.C=CC(=O)O.[Al]

Specifications

Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information soluble in acetone,benzene,carbon tetrachloride. Miscible with alcohol,ether,ethanol,ethyl ether,chloroform.
Formula Weight 243.17
Physical Form Powder
Chemical Name or Material Aluminum acrylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-836-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.