Learn More
Sigma Organic Chemistry Aluminum Doped Zinc Oxide Nanoparticle Ink | 10ML | 2185-0-0-1
Supplier:  Sigma Organic Chemistry 92156410ML
Encompass_Preferred
Aluminum Doped Zinc Oxide Nanoparticle Ink 2.5 Wt% Viscosity 3.5 Cp Work Function Variation 3.9 Ev, 10ml
About This Item:
Unspsc Code: 12352200
Storage: Room Temp Controlled
Catalog No. 92-156-410ML
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.