Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Brunswick Scientific™ Erlenmeyer Flask Dedicated Platform

Catalog No. 14278054 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Innova 44/44R Shakers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-278-054 Innova 44/44R Shakers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-278-054 Supplier New Brunswick Scientific™ Supplier No. M12829926
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use this heavy duty aluminum plate with Innova 44/44R shakers. New Brunswick Scientific™ Erlenmeyer Flask Dedicated Platforms are recommended for shaking full loads of flasks, 1L and larger, containing more than 20% volume or with baffles.

  • 2L Erlenmeyer flask dedicated platform
  • Heavy duty 0.95cm thick aluminum
  • For Innova 44/44R Incubator Shakers
  • 76 x 46cm
  • Not interchangeable with other 76 x 46cm platforms
TRUSTED_SUSTAINABILITY

Specifications

Product Type Erlenmeyer Flask Dedicated Platform
Material Aluminum
Dimensions (L x W) 76 x 46 cm
Thickness (Metric) 0.95 cm
For Use With (Equipment) Innova 44/44R Shakers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.