Learn More
New Brunswick Scientific™ Aluminum Erlenmeyer Flask Platform
Catalog No. 14278046 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Innova 44/44R Incubator Shaker
Catalog No. For Use With (Equipment)
14-278-046 Innova 44/44R Incubator Shaker
1 options
Catalog No. 14-278-046 Supplier New Brunswick Scientific™ Supplier No. M12829907
Add to Cart
Add to Cart
Use these trays with Innova™ 44/44R stackable incubator shakers. New Brunswick Scientific™ Aluminum Erlenmeyer Flask Platforms are designed for 500mL Erlenmeyer flasks
  • Aluminum Erlenmeyer flask dedicated platform
  • 76 × 46cm
  • Not interchangeable with other 76 × 46cm platforms
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Dimensions (L x W) 76 x 46 cm
For Use With (Equipment) Innova 44/44R Incubator Shaker