Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum ethoxide

Catalog No. AA4133618
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4133618 50 g
AA4133609 10 g
AA4133630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4133618 Supplier Thermo Scientific Chemicals Supplier No. 04133618
Add to Cart

CAS: 555-75-9 | C6H15AlO3-3 | 162.165 g/mol

Aluminum ethoxide is mainly used in industrial settings. It is used as a reducing agent for aldehyde, ketones, polymerization catalyst. Used in the sol-gel process preparation of high purity aluminum sesquioxide. It also used as a reducing reagent, for carbonyl compounds that restore to alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-75-9
Molecular Formula C6H15AlO3-3
Molecular Weight (g/mol) 162.165
MDL Number MFCD00009658
InChI Key JBRFPFQROJYWFN-UHFFFAOYSA-N
Synonym acmc-209lnt
PubChem CID 70700059
IUPAC Name aluminum;ethanolate
SMILES CC[O-].CC[O-].CC[O-].[Al]

Specifications

Melting Point 154°C to 159°C
Color White
Density 1.142 g/mL
Boiling Point 320°C
Linear Formula Al(OC2H5)3
Quantity 50 g
UN Number UN2925
Sensitivity Moisture sensitive
Merck Index 14,338
Solubility Information Soluble in hot xylene,cholorobenzene. Reacts with water.
Formula Weight 162.16
Physical Form Chunks and/or Powder
Chemical Name or Material Aluminum ethoxide
Show More Show Less
Hazard Category H228-H261-H314
Hazard Statement GHS H Statement
H228-H314
Flammable solid.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P223-P231+P232-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: II
EINECSNumber 209-105-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.