Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Fluoride, Anhydrous, 99.5% (metals basis)

Catalog No. AA3633514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3633514 25 g
AA3633506 5 g
AA3633522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3633514 Supplier Thermo Scientific Chemicals Supplier No. 03633514
Add to Cart

For vacuum deposition | CAS: 7784-18-1

Aluminum fluoride (AlF3) is an inorganic compound used primarily in the production of aluminum. It is also used to inhibit fermentation. It is a sputtering target for preparation of low index films. In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-18-1
MDL Number MFCD00003426

Specifications

Melting Point 1291°C (sublimation)
Physical Form Sintered Lump
Quantity 25 g
Merck Index 14,339
Solubility Information Sparingly soluble in acids and alkalies. Insoluble in Acetone.
Formula Weight 83.98
Percent Purity 99.5%
Sensitivity Hygroscopic
Density 2.882 g/mL
Chemical Name or Material Aluminum fluoride, Anhydrous
Hazard Category Danger
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 232-051-1
RTECSNumber BD0725000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.