Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum foil, 0.2mm (0.008in) thick, annealed, Puratronic, 99.9999% (metals basis)

Catalog No. p-7059284
Change view
Click to view available options
Quantity:
100 x 100 mm
100 x 500 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44330GH 100 x 100 mm
AA44330HP 100 x 500 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44330GH Supplier Thermo Scientific Chemicals Supplier No. 044330GH
Only null left
Add to Cart

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Quantity 100 x 100 mm
Odor Odorless
Weight ≈5.4g/100x100mm
Form Foil
IUPAC Name aluminium
Percent Purity 99.9999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Aluminum foil, 0.2mm (0.008 in.) thick, Annealed, Temper: soft
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.