Learn More
Grainger Aluminum Foil Roll 500 ft L 18in W Heavy
Supplier:  Grainger 16W483
Encompass_Preferred
The Aluminum Foil Roll Heavy Weight from Grainger is a commercial-grade aluminum foil roll ideal for food service and industrial applications. It measures 18 inches wide x 500 feet long and is constructed from heavy gauge aluminum for increased durability. The foil is designed for wrapping, lining, and masking applications. It can withstand temperatures up to 400°F. The foil roll allows for quick food coverage and provides an excellent barrier against moisture, oils, and odors. Overall, this heavy duty aluminum foil delivers the strength needed for commercial use.
Catalog No. 50-276-6349
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.