Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum iodide, 99.99+% (metals basis)

Catalog No. AA8918506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8918506 5 g
AA8918514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8918506 Supplier Thermo Scientific Chemicals Supplier No. 8918506
Add to Cart

Moisture and Light Sensitive | CAS: 7784-23-8 | AlI3 | 407.695 g/mol

Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-23-8
Molecular Formula AlI3
Molecular Weight (g/mol) 407.695
MDL Number MFCD00003427
InChI Key CECABOMBVQNBEC-UHFFFAOYSA-K
Synonym aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis
PubChem CID 82222
IUPAC Name triiodoalumane
SMILES [Al](I)(I)I

Specifications

Boiling Point 360°C
Melting Point 191°C
Color Brown-Purple to White
Physical Form Lump
Quantity 5 g
Assay Percent Range >99.99% (Metals basis)
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 407.69
Odor Pungent
Sensitivity Light and moisture sensitive
Density 3.98
Chemical Name or Material Aluminum iodide
Show More Show Less
Hazard Category H314-H335
Hazard Statement Keep container dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 232-054-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.