Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Aluminum Keyed Alike Safety Lock with Steel Shackle

Catalog No. 19000739 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
Yellow
Quantity:
12 Pk.
24 Pk.
6 Pk.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-000-739 Red 12 Pk.
19-000-733 Red 6 Pk.
19-000-734 Yellow 6 Pk.
19-000-735 Blue 6 Pk.
19-000-736 Green 6 Pk.
19-000-737 Orange 6 Pk.
19-000-741 Red 24 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-000-739 Supplier Brady™ Supplier No. SDALRED38STKA12
Add to Cart
Add to Cart
This item is not returnable. View return policy

Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient and cost-effective Safety Lockout Padlocks from Brady.

  • Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program.
  • Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
  • Aluminum padlocks with steel shackles are durable, corrosion-resistant and ideal for general lockout applications.
  • Locks that are Keyed Different (KD) are ideal for simple lockout applications and a manageable number of energy isolation points.
  • Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
  • Record and retain engraved key codes to streamline replacement of lost keys.

Specifications

Product Type Padlock
Key Type Keyed Alike
Color Red
Material Aluminum
Diameter (English) Shackle 0.244 in.
Diameter (Metric) Shackle 6.20 mm
Dimensions (D x W x H) 0.75 x 1.5 x 1.8 in.
Height (Metric) Shackle 38.10 mm
Includes (12) Padlocks, (24) Keys
Material (Shackle) Steel
No. of Pins 6
Quantity 12 Pk.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.