Learn More
Aluminum lactate, 90-95%, contains ^=5% water, Thermo Scientific Chemicals
Description
It is used as a skin moisturizer lotion to treat dry, scaly, itchy skin, reagent in biological activity studies of aluminum and in sol-gel processing. Also used in fire foam. Aluminum L-Lactate has been used to inject aluminum into rabbits. Aluminum L-Lactate has been shown to decrease human enamel dissolution in a pH 4 acetate buffer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 100 g |
Merck Index | 14,348 |
Solubility Information | Very soluble in cold water |
Formula Weight | 294.2 |
Percent Purity | 90 to 95% |
Concentration or Composition (by Analyte or Components) | Contains ≈5% water |
Odor | Odorless |
Chemical Name or Material | Aluminum lactate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 242-670-9 |
RTECSNumber | BD2214000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.