Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Metal, 200 Mesh, Powder, Spectrum™ Chemical
SDP

Catalog No. 18608524 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-524 500 g Poly Bottle
18-608-520 25 g Poly Bottle
18-608-522 100 lb. Poly Bottle
18-608-523 2.5 kg Poly Bottle
18-608-521 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-524 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1050500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7429-90-5

Specifications

CAS Max % 1
Quantity 500 g
Packaging Poly Bottle
Form Powder
Grade Ungraded
Chemical Name or Material Aluminum Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.