Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Aluminum Metal Strips
SCI_ED

Catalog No. S25146 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
30 Ea.
Packaging:
Lab Grade - Metal Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25146 30 Ea. Lab Grade - Metal Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25146

missing translation for 'validMainframeMsg'

Reacts with water or moisture

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Density 2.7g/mL
Quantity 30 Ea.
Packaging Lab Grade - Metal Strips
Linear Formula Al
Formula Weight 26.98
Chemical Name or Material Aluminum
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.