Learn More
Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis)
Description
Aluminum nitrate is a strong oxidizing agent. It is used in tanning leather, antiperspirants, corrosion inhibitors, extraction of uranium, petroleum refining, and as a nitrating agent. The nonahydrate and other hydrated aluminum nitrates have many applications. These salts are used to produce alumina for preparation of insulating papers, in cathode ray tube heating elements, and on transformer core laminates. The hydrated salts are also used for the extraction of actinide elements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 73.5°C (135° (decomposition)) |
| Physical Form | Crystalline |
| Quantity | 500 g |
| UN Number | UN1438 |
| Merck Index | 14,352 |
| Solubility Information | Soluble in water (540mg/ml),ethanol,alcohol (very),and acetone (slightly). |
| Formula Weight | 375.13 (212.99 Anhydrous) |
| Percent Purity | 99.999% |
| Grade | Puratronic |
| Odor | Odorless |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H318 |
| missing translation for 'healthHazard2' | GHS H Statement H272-H315-H319 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. |
| missing translation for 'healthHazard3' | P280-P305+P351+P338-P310 |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: ALUMINUM NITRATE |
| EINECSNumber | 236-751-8 |
| RTECSNumber | BD1050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.