Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Nitride Paste, Thermo Scientific™

Catalog No. AA4395936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4395936 500 g
AA4395914 25 g
AA4395922 100 g
AA43959A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4395936 Supplier Thermo Scientific Chemicals Supplier No. 4395936

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24304-00-5
Molecular Formula AlN
Molecular Weight (g/mol) 40.99
MDL Number MFCD00003429
InChI Key PIGFYZPCRLYGLF-UHFFFAOYSA-N
Synonym aluminum nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminum nitride grade a, aluminum nitride grade b, aluminum nitride grade c
PubChem CID 90455
ChEBI CHEBI:50884
IUPAC Name alumanylidyneamine
SMILES N#[Al]

Specifications

Color Gray
Physical Form Paste
Quantity 500 g
Odor Odorless
Appearance Gray paste
Density 1.98
Chemical Name or Material Aluminum Nitride, Air set in 1-4 hours, heat cure in 2 hours at 200°C
Hazard Category H315-H319-H372
Precautionary Statement P260-P264b-P270-P280-P302+P352-P305+P351+P338-P314-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 246-140-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.