Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxide Standard, MilliporeSigma™ Supelco™

Catalog No. 111022409 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2409 5 kg
11-102-2407 1 kg
11-102-2408 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2409 Supplier MilliporeSigma Supelco Supplier No. 063005KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration. Aluminum oxides are generally used as solid support phases in solid phase extraction (SPE)-based clean-up procedures employed in chromatographic methods.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Grade Standard
Melting Point 2040°C (literature)
Quantity 5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.