Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Pellets, Reagents

Catalog No. 01337510
Change view
Click to view available options
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-510 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-510 Supplier Reagents Holdings Llc Supplier No. C2123000500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aluminum Pellets, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym Aluminium
IUPAC Name aluminium
SMILES [Al]

Specifications

Color Silver-Gray
Melting Point 660°C
Boiling Point 2467°C
Avg. Mol. Wt. or Mol. Wt. Range 26.9815
Quantity 500 g
Packaging Poly Bottle
Form Solid
Chemical Name or Material Aluminum Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.