Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum phosphate, Puratronic™, 99.99% (metals basis)

Catalog No. p-4407201
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1062806 5 g
AA1062814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1062806 Supplier Thermo Scientific Chemicals Supplier No. 01062806
Only null left
Add to Cart

CAS: 7784-30-7 | AlO4P | 121.951 g/mol

Aluminum phosphate is used in the production of glass, mixture of ceramic, waterproofing concrete and dental cements, in cosmetics as a emollient, flame retardant, catalyst in organic synthesis, fireproofing textile, pharmaceuticals and dyes. It also used in many adhesives, binders and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-30-7
Molecular Formula AlO4P
Molecular Weight (g/mol) 121.951
MDL Number MFCD00003430
InChI Key ILRRQNADMUWWFW-UHFFFAOYSA-K
Synonym aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1
PubChem CID 64655
IUPAC Name aluminum;phosphate
SMILES [O-]P(=O)([O-])[O-].[Al+3]

Specifications

Melting Point >1500°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in concentrated hydrochloric acid,alcohol and nitric acid. Insoluble in water.
Formula Weight 121.95
Appearance White powder
Density 2.566 g/cm3
Chemical Name or Material Aluminum phosphate
Hazard Category H315-H320
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P315-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 232-056-9
RTECSNumber TB6450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.