Learn More
Medchemexpress LLC Aluminum phthalocyanine chloride | 14154-42-8 | MFCD00049386 | C32H16AlClN8 | 10MG

Supplier: Medchemexpress LLC HY12995910MG
Aluminum phthalocyanine chloride is a chloroaluminum phthalocyanine photosensitizer used in photodynamic research and photosensitization studies. It is supplied as a blue to dark-blue solid (C32H16AlClN8, CAS 14154-42-8). Purity is batch-specific; consult the COA and SDS for exact specifications and handling instructions.
- Used as a photosensitizer in photodynamic therapy and photosensitization research.
- Blue to dark-blue solid suitable for laboratory handling and formulation.
- Chemical formula C32H16AlClN8 and CAS 14154-42-8 documented for identification.
- Purity provided per batch via certificate of analysis.
- Store sealed, away from moisture and light; follow COA/SDS for solvent storage conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.