Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Aluminum platform for Innova™ 44/44R

Catalog No. 14278045 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
250 mL Erlenmeyer Flask Dedicated Platform
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-278-045 250 mL Erlenmeyer Flask Dedicated Platform
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-278-045 Supplier Eppendorf™ Supplier No. M12829906
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eppendorf shaker platforms are available in a wide variety of materials and sizes, perfect for virtually every requirement. Universal platforms can be adapted for thousands of combinations of clamps and other accessories. Dedicated platforms come complete with a single clamp size pre-installed.

Aluminum platform for Innova™ 44/44R, 76 × 46 cm, not interchangeable with other 76 × 46 cm platforms, 250 mL Erlenmeyer flask dedicated platform- Flask Capacity: 40
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 76 x 46 cm
Description Not Interchangeable With Other
For Use With (Equipment) 250 mL Erlenmeyer Flask Dedicated Platform
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.