Learn More
Selleck Chemical LLC 2-Mercaptobenzothiazole
SDP
Supplier:  Selleck Chemical LLC E01841G
Encompass_Preferred
2-Mercaptobenzothiazole (Benzothiazole-2-thiol 2-MBT Mercapto-2-benzothiazole) is an exceptionally potent inhibitor of banana polyphenoloxidase and can significantly delay the onset of substrate oxidation at concentrations as low as 100 nM
Catalog No. 50-313-7575
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.