Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis), Thermo Scientific Chemicals

Catalog No. AA41001A7
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41001A7 5 kg
AA41001A1 1 kg
AA4100130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41001A7 Supplier Thermo Scientific Chemicals Supplier No. 041001A7
Add to Cart

missing translation for 'validMainframeMsg'

Moisture Sensitive; Tap density 1.5g/cm3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Color Silver to Gray
Appearance Silver Gray powder
Quantity 5 kg
Odor Odorless
Sensitivity Water reactive
Form Powder
Assay Percent Range 98% (Metals basis)
Chemical Name or Material Aluminum, APS 10.0-14.0 micron
What is the tap density of Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?

The tap density of Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 1.4.

What is the iron wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?

The iron wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 0.2?

What is the oil and grease wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?

The oil and grease wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 0.2.

What is the volatile wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?

The volatile wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 0.1.

What is the free metallic aluminum wt% minimum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?

The free metallic aluminum wt% minimum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 97.5.

Hazard Category H250-H261-H500
Precautionary Statement P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-072-3
RTECSNumber BD0330000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.