Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum rod, 9.5mm (0.37in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number AA10779CZ
Click to view available options
Quantity:
5 cm
25 cm
4 x 25 cm
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
aluminum
[Al]
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
aluminum
[Al]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 4 x 25 cm
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Weight ≈1.91g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Aluminum rod, 9.5mm (0.37 in.) dia.
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit