Learn More
Labconco Aluminum Rotor with Pear Shaped Flasks for CentriVap™ Pro
GREENER_CHOICESDP
Catalog No. 12199550 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
PTFE-coated Stainless Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
12-199-550 Stainless Steel
12-199-548 PTFE-coated Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-199-550 Supplier Labconco Supplier No. 7556600
Add to Cart
Add to Cart
Rotor for use with Labconco™ CentriVap™ Pro Vacuum Concentrators.
  • Compatible with CentriVap™ Pro Vacuum Concentrators
Specifications
Material Stainless Steel
Type Rotor
Holds 100 mL Pear Shaped Flasks