Learn More
Brady™ Aluminum Sign: DANGER - CYLINDERS CONTAIN EXPLOSIVE. GAS HANDLE WITH CARE.
Catalog No. 19105699 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
Catalog No. Dimensions (L x W)
19-105-699 10 x 7 in.
1 options
Catalog No. 19-105-699 Supplier Brady™ Supplier No. 126186
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Aluminum
Shape Rectangular
Dimensions (L x W) 10 x 7 in.
Displayed Text DANGER - CYLINDERS CONTAIN EXPLOSIVE. GAS HANDLE WITH CARE.
Color Black
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.