Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Sulfate X-hydrate, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S000522 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000522 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger: Causes serious eye damage.
Catalog No. S000522 Supplier Aldon Corporation Supplier No. IS10132
Add to Cart
Add to Cart
Danger: Causes serious eye damage.

Chemical Identifiers

CAS 16828-12-9
Molecular Weight (g/mol) ∼594 g/mol

Specifications

Physical Form Flakes or Powder
Quantity 500 g
Percent Purity 100%
Grade Laboratory
Packaging HDPE
Chemical Name or Material Aluminum Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.