Learn More
United Scientific™ Aluminum Weighing Dishes, Round
SDPSCI_ED
Catalog No. S80002 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Diameter (Metric):
38.1 mm
71.1 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S80002 38.1 mm
S80003 71.1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S80002 Supplier United Scientific™ Supplier No. UNWGBTAL4PK50
Only null left
Add to Cart
Add to Cart
Weighing Dishes are ideal for general purpose weighing, dispensing, storing, sampling, and testing applications.
  • These smooth-wall, flat-bottom aluminum dishes are ideal for general purpose weighing, dispensing, storing, sampling, and testing applications.
  • Dishes include tapered sides to enable easy stacking.
  • Ideal for general-purpose applications
  • Slightly tapered sides for easy stacking
  • Disposable
  • Aluminum construction
Specifications
Autoclavable Not Autoclavable
Bottom Shape Flat
Color White
Disposable Single-use
Material Polystyrene
Shape Circular
Sterility Non-sterile
Type Weighing Dish
Diameter (Metric) 38.1 mm