Learn More
Iwai North America ALyS505N-7: Xeno-free medium with IL-2 (700IU/mL) for activation of human lymphocytes. Size: 1L
Supplier:  Iwai North America 1027P10
Encompass_Preferred
ALyS505N-7 is ALyS505N with IL-2 (700IU/mL) is Xeno-free medium suitable for activating T cells from human lymphocytes. Higher concentrations of IL-2 activate T cells, while lower concentrations are utilized for expansion.1. Futami, M. et al. The novel multi-cytokine inhibitor TO-207 specifically inhibits pro-inflammatory cytokine secretion in monocytes without affecting the killing ability of CAR T cells. PLOS ONE 15, e0231896 (2020) DOI: 10.1371/journal.pone.0231896
Catalog No. NC2733018
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.