Learn More
Cellular Engineering Technologies Cryopreserved vial of induced pluripotent stem cells from a single male donor with Alzheimer's Presenilin-1 Mutation (500,000 cells)
Supplier:  Cellular Engineering Technologies CR1008500
Encompass_Preferred
Cell line was derived from a single male with Alzheimer's disease, with onset occurring at the age of 45. The donor cells were collected via skin biopsy of the forearm antemortem. The cell morphology is fibroblast-like. Vial contains 500,000 cells. Shipped with dry ice.Mutations in the PSEN1 gene, encoding presenilin-1 (PS1), are the most common common cause of familial Alzheimer's disease (FAD). PS1 is the catalytic subunit of y-secretase, an intramembranous protease that cleaves various type 1 transmembrane proteins, notably the amyloid precursor protein (APP) and Notch.Vial contains approximately 500,000 cells. Shipped with dry ice.
Catalog No. NC2753013
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.