Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AM 251

Catalog No. p-4783242
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-171 1 mg
11-171-0 10 mg
11-175-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-171 Supplier Tocris Bioscience™ Supplier No. 1117/1
This item is not returnable. View return policy

Potent CB1 antagonist; also GPR55 agonist

Potent CB1 receptor antagonist (IC50 = 8 nM, Ki = 7.49 nM) that displays 306-fold selectivity over CB2 receptors. Also potent GPR55 agonist (EC50 = 39 nM) and μ-opioid receptor antagonist (Ki = 251 nM).

Fluorescent Form also available.

Chemical Identifiers

CAS 183232-66-8
Molecular Formula C22H21Cl2IN4O

Specifications

Quantity 1 mg
Formula Weight 555.24
Purity >99%
Molecular Weight (g/mol) 555.241
Chemical Name or Material AM 251
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.