Learn More
Medchemexpress LLC 4-[4-[1-(2,4-dichlorophenyl)-4-methyl-3-(piperidinylcarbamoyl)pyrazol-5-yl]phenyl]but-3-ynyl nitrate | 1245626-00-9 | 99.9% | 100 MG
Supplier: Medchemexpress LLC HY120423100MG
AM-6538 is a long-acting, high-affinity, pseudo-irreversible cannabinoid (CB) receptor antagonist used in research to evaluate the apparent efficacy of cannabinoid agonists. It is a structural analog of rimonabant and contains an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry.
- Long-acting, high-affinity cannabinoid receptor antagonist.
- Pseudo-irreversible binding useful for receptor availability studies.
- Alkyne functional group compatible with copper-catalyzed azide-alkyne cycloaddition.
- High purity: 99.9%.
- Available as a 100 mg solid.