Learn More
Medchemexpress LLC 4-[4-[1-(2,4-dichlorophenyl)-4-methyl-3-(piperidinylcarbamoyl)pyrazol-5-yl]phenyl]but-3-ynyl nitrate | 1245626-00-9 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12042325MG
Encompass_Preferred
AM-6538 is a long-acting, high-affinity, pseudo-irreversible cannabinoid CB1 receptor antagonist supplied as a solid powder for research use. It is a structural analog of rimonabant and is commonly used in preclinical receptor pharmacology, ligand-binding stabilization, and structural studies.
  • Long-acting CB1 receptor antagonist.
  • High affinity and pseudo-irreversible binding.
  • Supplied as a solid powder with high purity.
  • Suitable for in vitro and preclinical pharmacology studies.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-34553
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.