Learn More
Abcam AM 80, RAR Alpha/Beta Agonist, Solid, 10 mg
Description
- Purity: >98%
- Molecular Weight: 351.44 g/mol
- Activity: Selective retinoic acid receptor α/β (RARα/β) agonist
- Biological Effect: Induces differentiation and anti-cancer activity in vivo and in vitro
- Form: Solid
- CAS Number: 94497-51-5
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% AM 80 |
| Target Molecule | Retinoic Acid Receptor alpha/beta |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Formulation | 100% AM 80 |
| Quantity | 10 mg |
| For Use With (Application) | Induces differentiation and anti-cancer activity |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.